Vanadium(V)oxid ≥99,9% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

44424.22
44424.22EA 944 EUR
44424.22
Vanadium(V)oxid ≥99,9% (Metall-Basis)
Vanadium(V)oxid

Purity: 99.9% CAS No.: 1314-62-1 Molecular Formula: V2O5 Formula Weight: 181.88 Physical Form: 3-12mm Pieces Density: 3.357 Melting Point: 690° Boiling Point: 1750° dec. Merck Reference: 14,9921 UN No.: 2862 MDL No.: MFCD00011457


Vacuum deposition

Formel: O₅V₂
Siedepunkt: 1750 °C (Zersetzung)
Schmelzpunkt: 690 °C
Dichte: 3,357
Lagertemperatur: Raumtemperatur
MDL: MFCD00011457
CAS-Nummer: 1314-62-1
EINECS: 215-239-8
UN: 2862
ADR: 6.1,III
Merck Index: 14,9921

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR