MMP-9/MMP-13 inhibitor I ≥95% (durch HPLC), Millipore®
Lieferant: Merck Millipore Calbiochem
Verpackt unter Schutzgas
444252-1
444252-1EA
131
EUR
444252-1
MMP-9/MMP-13 inhibitor I ≥95% (durch HPLC), Millipore®
MMP-9/MMP-13 inhibitor I
A piperazine-based, cell-permeable, and highly potent inhibitor of MMP-9 (IC₅₀=900 pM) and MMP-13 (IC₅₀=900 pM).
Inhibits MMP-1 and MMP-3 at much higher concentrations (IC₅₀=43 nM and 23 nM, respectively). Also acts as an inhibitor of MMP-7 (IC₅₀=930 nM).
Formel:
C₂₅H₂₅N₃O₆S Lagertemperatur: Gefrierschrank |
MDL:
MFCD03453559 CAS-Nummer: 204140-01-2 |
Spezifikation Testergebnisse
Purity (HPLC) | ≥95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...