Metanilgelb, Supelco®

Lieferant: Merck

Synonyme: Acid Yellow 36, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt

M-Clarity™ Qualitätsstufe = MQ100

44426-100MG
44426-100MGEA 265 EUR
44426-100MG
Metanilgelb, Supelco®
Metanilgelb
Formel: C₁₈H₁₄N₃NaO₃S
Molecular Weight: 375,38 g/mol
MDL: MFCD00007486
CAS-Nummer: 587-98-4
EINECS: 209-608-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4ba3794ad6f77ec228ae5f753be341681ea169e&vid=ff9aa10a234c2076d66fac301e3bf6dcd8e746da&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR