Chloroform ≥99,5% stabilisiert, AR®, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr


Stabilisierung: Stabilisiert mit Ethanol ~1%

4444-45EA 718.69 EUR
4444-45 4444-25
Chloroform ≥99,5% stabilisiert, AR®, Macron Fine Chemicals™
Chloroform
Formel: CHCl₃
MDL: MFCD00000826
CAS-Nummer: 67-66-3
UN: 1888
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use ≥99.5%
Chloride Ion and Free Chlorine Passes Test
Nonvolatile Residue (w/v) ≤0.0014 %
Water (H₂O) Passes Test
Specific Gravity at 25°/25°C 1.474 - 1.482
Appearance (clear, colorless liquid) Passes test
Free Chlorine (Cl2) ≤0.0005 %
Stabiliser (Ethanol) ~1%
For Laboratory, Research or Manufacturing Use


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR