Mangan(II)oxid ≥99,99% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Manganmonoxid, Manganoxid

44441.09 44441.18
44441.09EA 159 EUR
44441.09 44441.18
Mangan(II)oxid ≥99,99% (Metall-Basis)
Mangan(II)oxid

Purity: 99.99% CAS No.: 1344-43-0 Molecular Formula: MnO Formula Weight: 70.94 Physical Form: Powder/Chunks Density: 5.0 Appearance: Green Odour: Odourless MDL No.: MFCD00016226

Formel: MnO
Schmelzpunkt: 1840 °C
Dichte: 5,0
Lagertemperatur: Raumtemperatur
MDL: MFCD00016226
CAS-Nummer: 1344-43-0
EINECS: 215-695-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR