Certribromid 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Cer(III)bromid

44447.14 44447.22
44447.14EA 126 EUR
44447.14 44447.22
Certribromid 99%
Certribromid

Purity: 99% CAS No.: 14457-87-5 Molecular Formula: CeBr3.xH2O Formula Weight: 379.83(anhy) Physical Form: Crystalline Odour: Odourless MDL No.: MFCD00150206

Formel: CeBr₃
Molecular Weight: 379.83 g/mol
Schmelzpunkt: 732
Lagertemperatur: Raumtemperatur
MDL: MFCD00016004
CAS-Nummer: 14457-87-5
EINECS: 238-447-0
Merck Index: 12,02039

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR