Cerium(IV) isopropoxide (37 - 45% Ce)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
CAS No.: 63007-83-0
Molecular Formula: C12H28CeO4
Formula Weight: 376.48
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
Appearance: Orange-red to yellow
Melting Point: 140° dec.
Solubility: Soluble in pyridine
UN No.: UN2925
MDL No.: MFCD00061483
Formel:
C₁₂H₂₈CeO₄ Schmelzpunkt: 140 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061483 CAS-Nummer: 63007-83-0 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...