Hexaamminnickel(II)bromid ≥99,999% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
44482.14
44482.14EA
98.1
EUR
44482.14
Hexaamminnickel(II)bromid ≥99,999% (Metall-Basis)
Hexaamminenickel(II) bromide
Purity: 99.999%
CAS No.: 13601-55-3
Molecular Formula: Ni(NH3)6Br2
Formula Weight: 320.68
Physical Form: Crystalline
Formel:
H₁₈Br₂N₆Ni Molecular Weight: 320.68 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039935 CAS-Nummer: 13601-55-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...