2-(Hydroxymethyl)-15-krone-5 ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(Hydroxymethyl)-15-crown-5-ether

44500.03 44500.06
44500.03EA 104 EUR
44500.03 44500.06
2-(Hydroxymethyl)-15-krone-5 ≥98%
2-(Hydroxymethyl)-15-krone-5

Purity: 98% CAS No.: 75507-25-4 Molecular Formula: C11H22O6 Formula Weight: 250.29 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.175 Refractive Index: 1.479 Boiling Point: 135°/1.5mm Beilstein Reference: 3589706 MDL No.: MFCD00192215

Formel: C₁₁H₂₂O₆
Molecular Weight: 250.29 g/mol
Siedepunkt: 135 °C (1.5 mmHg)
Dichte: 1.175
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00192215
CAS-Nummer: 75507-25-4
EINECS: 278-222-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR