Fullerene buckytube/nanotube einwandig >60%

Lieferant: Thermo Scientific
Achtung

44501.02
44501.02EA 597 EUR
44501.02
Fullerene buckytube/nanotube einwandig >60%
Fullerene buckytube/nanotube

CAS No.: 308068-56-6 Physical Form: Powder Appearance: Black UN No.: UN1325 MDL No.: MFCD00282904

Lagertemperatur: Raumtemperatur
MDL: MFCD00282904
CAS-Nummer: 308068-56-6
UN: 1325
ADR: 4.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR