Poly(styrene), Testfolie
Lieferant: Thermo Fisher Scientific
44502.KT
44502.KTEA
75.8
EUR
44502.KT
Poly(styrene), Testfolie
Poly(styrene)
Formel:
C₈H₈ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084450 CAS-Nummer: 9003-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...