Platin(IV)chlorid ≥99,99% (Metall-Basis) Pt ≥ 57%, Premion®

Lieferant: Thermo Scientific
Gefahr

Synonyme: Platintetrachlorid

44503.06
44503.06EA 605 EUR
44503.06
Platin(IV)chlorid ≥99,99% (Metall-Basis) Pt ≥ 57%, Premion®
Platin(IV)chlorid

Purity: 99.99+% CAS No.: 13454-96-1 Molecular Formula: PtCl4 Formula Weight: 336.90 Physical Form: Crystalline Soluble Density: 4.303 Appearance: Brown Odour: Odourless Melting Point: 370° dec. UN No.: 2923 MDL No.: MFCD00011182

Formel: Cl₄Pt
Molecular Weight: 336,89 g/mol
Schmelzpunkt: 370 °C (Zers.)
Dichte: 4,303
Lagertemperatur: Raumtemperatur
MDL: MFCD00011182
CAS-Nummer: 13454-96-1
EINECS: 236-645-1
UN: 2923
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR