2-(Hydroxymethyl)-12-krone-4 ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

44509.01 44509.03 44509.06
44509.01EA 42.9 EUR
44509.01 44509.03 44509.06
2-(Hydroxymethyl)-12-krone-4 ≥97%
2-(Hydroxymethyl)-12-krone-4

Purity: 97% CAS No.: 75507-26-5 Molecular Formula: C9H18O5 Formula Weight: 206.24 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.20 Refractive Index: 1.4800 Boiling Point: 115°/0.04mm Beilstein Reference: 4661968 MDL No.: MFCD00067524

Formel: C₉H₁₈O₅
Siedepunkt: 115 °C (0,04 mmHg)
Dichte: 1,20
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00067524
CAS-Nummer: 75507-26-5
EINECS: 278-224-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR