n-Heptadecan ≥99,5%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
44514.03
44514.03EA
25
EUR
44514.03
n-Heptadecan ≥99,5%
n-Heptadecan
Purity: 99.5%
CAS No.: 629-78-7
Molecular Formula: CH3(CH2)15CH3
Formula Weight: 240.48
Physical Form: Crystalline
Density: 0.7783
Refractive Index: 1.4369
Melting Point: 22°
Boiling Point: 302°
MDL No.: MFCD00009002
Formel:
H₃C(CH₂)₁₅CH₃ Siedepunkt: 302 °C Schmelzpunkt: 22 °C Dichte: 0,7783 Flammpunkt: 148 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009002 CAS-Nummer: 629-78-7 EINECS: 211-108-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...