Titan(IV)oxid 20 - 35% in Wasser, kolloidale Dispersion

Lieferant: Thermo Scientific
Achtung

Synonyme: Pigment White 6, Titansäureanhydrid, Titandioxid

44517.A1
44517.A1EA 92.2 EUR
44517.A1
Titan(IV)oxid 20 - 35% in Wasser, kolloidale Dispersion
Titan(IV)oxid

CAS No.: 13463-67-7 Molecular Formula: TiO2 Formula Weight: 79.90 Density: 1.26 Boiling Point: 368° MDL No.: MFCD00011269


Applications: UV protection in clear coatings

Formel: TiO₂
Molecular Weight: 79,88 g/mol
Siedepunkt: 368 °C
Dichte: 1,26
Lagertemperatur: Raumtemperatur
MDL: MFCD00011269
CAS-Nummer: 13463-67-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR