Kobalt ≥99,995% (Metall-Basis), Drahtform, Puratronic®, geglüht, Ø 1.5 mm (0.06 in)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
44525.BS
44525.BW
44525.BSEA
191
EUR
44525.BS
44525.BW
Kobalt ≥99,995% (Metall-Basis), Drahtform, Puratronic®, geglüht, Ø 1.5 mm (0.06 in)
Cobalt
Purity: 99.995%
CAS No.: 7440-48-4
Physical Form: ≈15.8g/m
Odour: Odourless
MDL No.: MFCD00010935
Formel:
Co Siedepunkt: 2870 °C (1013 hPa) Schmelzpunkt: 1495 °C Dichte: 8,9 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010935 CAS-Nummer: 7440-48-4 EINECS: 231-158-0 Merck Index: 12,02488 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...