Phenol ≥99,5% Unstabilisiert
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Unstabilized
Purity: 99.5%
CAS No.: 108-95-2
Molecular Formula: C6H5OH
Formula Weight: 94.11
Flash Point: 79°(175°F)
Physical Form: Solid melt
Density: 1.071
Sensitivity: Hygroscopic
Odour: Sweet
Melting Point: 40-43°
Boiling Point: 181°
Beilstein Reference: 969616
Merck Reference: 14,7241
UN No.: 1671
MDL No.: MFCD00002143
Molecular biology
Formel:
C₆H₅OH Siedepunkt: 181 °C Schmelzpunkt: 40…43 °C Dichte: 1,071 Flammpunkt: 79 °C (175 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002143 CAS-Nummer: 108-95-2 EINECS: 203-632-7 UN: 1671 ADR: 6.1,II REACH: 01-2119471329-32 Merck Index: 14,7241 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...