Poly(styrene) ataktisch, M.W. 125,000-250,000
Lieferant: Thermo Fisher Scientific
44537.09
44537.18
44537.09EA
36.9
EUR
44537.09
44537.18
Poly(styrene) ataktisch, M.W. 125,000-250,000
Poly(styrene)
CAS No.: 9003-53-6
Formula Weight: 125,000-250,000
Physical Form: Beads
Odour: Odourless
MDL No.: MFCD00084450
Formel:
C₈H₈ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084450 CAS-Nummer: 9003-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...