Chrom ≥99,996% (Metall-Basis), Klumpen 2-15 mm (0.08-0.59 in)

Lieferant: Thermo Fisher Scientific

44565.09 44565.18
44565.09EA 99 EUR
44565.09 44565.18
Chrom ≥99,996% (Metall-Basis), Klumpen 2-15 mm (0.08-0.59 in)
Chromium

Purity: 99.996% CAS No.: 7440-47-3 Odour: Odourless MDL No.: MFCD00010944

Formel: Cr
Molecular Weight: 52 g/mol
Siedepunkt: 2469
Schmelzpunkt: 1840
Dichte: 7.2 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010944
CAS-Nummer: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR