Chromium etchant
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
44582.WY
44582.WYEA
107
EUR
44582.WY
Chromium etchant
Chromium etchant
Physical Form: Liquid
Density: 1.16
Appearance: Orange
Odour: Acrid
UN No.: UN3093
MDL No.: MFCD07370678
Applications: High purity ceric ammonium nitrate system for precise, clean etching of chromium and chromium oxide films
Dichte:
1,16 Lagertemperatur: Raumtemperatur |
MDL:
MFCD07370678 UN: 3093 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...