Copper etchant
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
44583.XC
44583.WY
44583.XCEA
328
EUR
44583.XC
44583.WY
Copper etchant
Copper etchant
Applications: Iron (III) chloride solution for immersion etching copper, copper alloys and Kovar in printed circuit board applications
Physical Form: Liquid
Density: 1.19
Appearance: Dark yellow to brown
Odour: Acidic
UN No.: UN3264
MDL No.: MFCD00161912
Applications: Iron (III) chloride solution for immersion etching copper, copper alloys and Kovar in printed circuit board applications
Dichte:
1,19 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00161912 UN: 3264 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...