Yttrium(III) trifluoroacetate hydrate ≥99,9%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
44597.09
44597.18
44597.09EA
72.6
EUR
44597.09
44597.18
Yttrium(III) trifluoroacetate hydrate ≥99,9%
Yttrium(III) trifluoroacetate hydrate
Purity: 99.9%
CAS No.: 304851-95-4
Molecular Formula: Y(OOCCF3)3.xH2O
Formula Weight: 427.95(anhy)
Physical Form: Crystalline
MDL No.: MFCD05663780
Formel:
C₆F₉O₆Y·xH₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD05663780 CAS-Nummer: 304851-95-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...