(S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridin
Lieferant: Thermo Fisher Scientific
Synonyme:
CTH-(S)-Xylyl-P-Phos, (S)-4,4'-Bis(bis(3,5-dimethylphenyl)phosphino)-2,2',6,6'-tetramethoxy-3,3'-bipyridine
unter Argon abgefüllt
44614.02
44614.77
44614.02EA
286
EUR
44614.02
44614.77
(S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridin
(S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridin
CAS No.: 443347-10-2
Molecular Formula: C46H50N2O4P2
Formula Weight: 756.86
Physical Form: Powder, packaged under argon
Melting Point: 158-162°
MDL No.: MFCD04974235
Asymmetric hydrogenation
Formel:
C₄₆H₅₀N₂O₄P₂ Schmelzpunkt: 158…162 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04974235 CAS-Nummer: 443347-10-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...