N,N-Dimethylformamiddimethylacetal ∼ 97%, AcroSeal®
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1,1-Dimethoxytrimethylamin
446151000
446151000.EA
159
EUR
446151000.
N,N-Dimethylformamiddimethylacetal ∼ 97%, AcroSeal®
N,N-Dimethylformamiddimethylacetal
Formel:
C₅H₁₃NO₂ Molecular Weight: 119,16 g/mol Siedepunkt: 102…104 °C (760 mmHg) Dichte: 0,89 g/cm³ Flammpunkt: 7 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008482 CAS-Nummer: 4637-24-5 UN: 1993 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3950 to 1.3980 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...