Silicon(IV) 2-ethylhexanoate ≥75%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
44635.30
44635.30EA
606
EUR
44635.30
Silicon(IV) 2-ethylhexanoate ≥75%
Silicon(IV) 2-ethylhexanoate
CAS No.: 67939-81-5
Molecular Formula: C32H60O8Si
Formula Weight: 600.91
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Appearance: Yellow
UN No.: UN1170
MDL No.: MFCD00269842
Formel:
C₃₂H₆₀O₈Si Lagertemperatur: Raumtemperatur |
MDL:
MFCD00269842 CAS-Nummer: 67939-81-5 UN: 1170 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...