Silicon surface sensitizer, gold chloride 0.5%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
44637.MQ
44637.MQEA
0
EUR
44637.MQ
Silicon surface sensitizer, gold chloride 0.5%
Silicon surface sensitizer, gold chloride 0.5%
MDL No.: MFCD06412431
Applications: Used for pretreatment of planar silicon devices when plating with Nickel plating solution products 44069 and 44070.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...