ICP-MS interne Standardlösung, Specpure®

Lieferant: Thermo Fisher Scientific

44651.AE
44651.AEEA 184 EUR
44651.AE
ICP-MS interne Standardlösung, Specpure®
Standards Kationischer Standards Mehrelement-Standards

UN Nr.: UN3264
MDL Nr.: MFCD00803321


Konzentration:
Bi 100 µg/ml ±2%;
Ga 100 µg/ml ±2%;
In 100 µg/ml ±2%;
Sc 100 µg/ml ±2%;
Tb 100 µg/ml ±2%;
Y 100 µg/ml ±2%;
6Li 100 µg/ml ±2%

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR