Cer(IV)oxid ≥99,5% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99.5%
CAS No.: 1306-38-3
Molecular Formula: CeO2
Formula Weight: 172.12
Physical Form: 0.07-0.1 Micron APS Powder, S. A. 8-12m2/g
Density: 7.132
Appearance: Pale yellow
Odour: Odourless
Melting Point: 2600°
Merck Reference: 14,1989
MDL No.: MFCD00010933
Formel:
CeO₂ Siedepunkt: 3468 °C (1013 hPa) Schmelzpunkt: 2600 °C Dichte: 7,132 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010933 CAS-Nummer: 1306-38-3 EINECS: 215-150-4 Merck Index: 14,1989 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...