Erbium(III)oxid ≥99,5% (REO, Basis der Oxide der Metalle der seltenen Erden), Nano-Pulver
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Erbiumoxid, Dierbiumtrioxid
44668.09
44668.09EA
90.1
EUR
44668.09
Erbium(III)oxid ≥99,5% (REO, Basis der Oxide der Metalle der seltenen Erden), Nano-Pulver
Erbium(III)oxid
Purity: 99.5%
CAS No.: 12061-16-4
Molecular Formula: Er2O3
Formula Weight: 382.56
Physical Form: 40-55nm APS Powder, S. A. 13-17m2/g
Density: 8.64
Melting Point: 2400°
Solubility: Soluble in acids
Merck Reference: 14,3646
MDL No.: MFCD00010991
Formel:
Er₂O₃ Molecular Weight: 382,52 g/mol Schmelzpunkt: 2400 °C Dichte: 8,64 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010991 CAS-Nummer: 12061-16-4 EINECS: 235-045-7 Merck Index: 14,3646 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...