Poly(acrylic acid) 25% (w/w) in Wasser
Lieferant: Thermo Fisher Scientific
44669.22
44669.36
44669.22EA
63.3
EUR
44669.22
44669.36
Poly(acrylic acid) 25% (w/w) in Wasser
Poly(acrylic acid)
CAS No.: 9003-01-4
Molecular Formula: [-CH2CH(CO2H)-]
Physical Form: Liquid
Density: 1.09
MDL No.: MFCD00084394
Formel:
C₃H₄O₂ Dichte: 1,09 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084394 CAS-Nummer: 9003-01-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...