Ethyl hydrogen malonate ≥90%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Mono-Ethyl malonate, 3-Ethoxy-3-oxopropanoic acid

44692.06 44692.14 44692.22 44692.36
44692.06EA 37.7 EUR
44692.06 44692.14 44692.22 44692.36
Ethyl hydrogen malonate ≥90%
Ethyl hydrogen malonate

Purity: 90+% CAS No.: 1071-46-1 Molecular Formula: C5H8O4 Formula Weight: 132.12 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.119 Refractive Index: 1.4295 Appearance: Pale yellow Boiling Point: 106°/3mm Beilstein Reference: 1758845 MDL No.: MFCD00020490

Formel: C₅H₈O₄
Molecular Weight: 132.115 g/mol
Siedepunkt: 106 °C (3 mmHg)
Dichte: 1.119
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00020490
CAS-Nummer: 1071-46-1
EINECS: 213-992-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR