Palladium nickel, Drahtgeflecht 50 mesh woven from 0.1578mm (0.0062in)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
44698.FL
44698.GT
44698.FLEA
501
EUR
44698.FL
44698.GT
Palladium nickel, Drahtgeflecht 50 mesh woven from 0.1578mm (0.0062in)
Palladium nickel
Molecular Formula: Pd:Ni; 95:5 wt%
Physical Form: Wire Cloth
MDL No.: MFCD05863544
Applications: Filtration
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...