Poly(styrene-divinylbenzene), 2% quervernetzt mit Divinylbenzol 200-400 mesh
Lieferant: Thermo Fisher Scientific
Synonyme:
SDVB resin, Styrene-divinylbenzene resin, Styrene-divinylbenzene copolymer, Poly(styrene-co-divinylbenzene), Polystyrene crosslinked with divinylbenzene
44708.14
44708.22
44708.36
44708.14EA
59.6
EUR
44708.14
44708.22
44708.36
Poly(styrene-divinylbenzene), 2% quervernetzt mit Divinylbenzol 200-400 mesh
Poly(styrene-divinylbenzene)
CAS No.: 9003-70-7
Physical Form: Powder
Odour: Odourless
MDL No.: MFCD00146442
Applications: Preparation of Merrifield type resins
Formel:
[-CH₂CH(C₆H₅)-]x[-CH₂CH[C₆H₄(CHCH₂)]-]y Molecular Weight: 234.342 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00146442 CAS-Nummer: 9003-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...