ICP-MS interference check standard solution B for 6020, Specpure®
Lieferant: Thermo Fisher Scientific
Matrix: 5% HNO₃
UN No.: UN3264
MDL No.: MFCD00803324
Note: To be diluted 1:500 to achieve prescribed concentrations.
Concentration: Ag 10 µg/ml ±2%; Al 10 µg/ml ±2%; As 10 µg/ml ±2%; Cd 10 µg/ml ±2%; Co 10 µg/ml ±2%; Cr 10 µg/ml ±2%; Cu 10 µg/ml ±2%; Mn 10 µg/ml ±2%; Ni 10 µg/ml ±2%; Se 10 µg/ml ±2%; V 10 µg/ml ±2%; Zn 10 µg/ml ±2%
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...