Poly(3-hexylthiophene-2,5-diyl) regioregulär metallarm
Lieferant: Thermo Fisher Scientific
|
Achtung
|
44745.02
44745.77
44745.02EA
494
EUR
44745.02
44745.77
Poly(3-hexylthiophene-2,5-diyl) regioregulär metallarm
Poly(3-hexylthiophene-2,5-diyl)
CAS No.: 104934-50-1
Physical Form: Powder
Solubility: Soluble in Chloroform and tetrahydrofuran
MDL No.: MFCD00217686
Formel:
C₁₂H₂₀S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00217686 CAS-Nummer: 104934-50-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...