2-Fluorisonicotinsäure ≥98%
Lieferant: Thermo Scientific
Synonyme:
2-Fluoropyridine-4-carboxylic acid
44755.03
44755.03EA
174
EUR
44755.03
2-Fluorisonicotinsäure ≥98%
2-Fluorisonicotinsäure
Purity: 98%
CAS No.: 402-65-3
Molecular Formula: C6H4FNO2
Formula Weight: 141.10
Physical Form: Powder
Melting Point: 195-497°
MDL No.: MFCD02181194
Formel:
C₆H₄FNO₂ Molecular Weight: 141,10 g/mol Schmelzpunkt: 195…497 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02181194 CAS-Nummer: 402-65-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...