ICP-MS internal standard solution for 200,8 & CLP ILM05,2, Specpure®
Lieferant: Thermo Fisher Scientific
44789.AE
44789.AEEA
175
EUR
44789.AE
ICP-MS internal standard solution for 200,8 & CLP ILM05,2, Specpure®
Standards
Kationischer Standards
Mehrelement-Standards
These standards are applicable for the determination of elements as dissolved elements in ground waters, surface waters, and drinking water, and total recoverable elements in these waters as well as wastewaters, sludges, and soils samples.
Matrix: 5% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD03792350
Concentration: Bi 10 µg/ml ±2%; In 10 µg/ml ±2%; Lu 10 µg/ml ±2%; Sc 10 µg/ml; ±2%; Tb 10 µg/ml ±2%; Y 10 µg/ml ±2%; ₆Li 10 µg/ml ±2%
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...