Ammonium metatungstate hydrate
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Ammonium meta-tungsten oxide hydrate
Ammonium tungsten oxide hydrate is widely utilized as a raw material for the synthesis of tungsten catalysts, which is used in a variety of reactions like oxidation, hydroxylation, hydrogenation and polymerization. It is used as a reagent for chemical analysis like medical diagnosis. It is used as a precursor material for the preparation of high-purity WO3 which is used as thin-film substrate for certain semiconductor devices. It acts as a precursor to a variety of POM (polyoxometalates).
Incompatible with strong oxidizing agents.
CAS No.: 12028-48-7
Molecular Formula: (NH4)6W12O39.xH2O
Formula Weight: 2938.41(anhy)
Physical Form: Powder
Solubility: Soluble in water. Insoluble in alcohol
MDL No.: MFCD00042673
Formel:
(NH₄)₆W₁₂O₃₉·xH₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042673 CAS-Nummer: 12028-48-7 EINECS: 234-733-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...