(Perfluorocyclohexyl)methyl acrylate ≥96% stabilisiert

Lieferant: Thermo Fisher Scientific


Stabilisierung: Stabilisiert mit 4-Methoxyphenol (MEHQ) 50 ppm-100 ppm

44793.06
44793.06EA 88 EUR
44793.06
(Perfluorocyclohexyl)methyl acrylate ≥96% stabilisiert
(Perfluorocyclohexyl)methyl acrylate

Stab. with 100ppm 4-methoxyphenol


Purity: 96% CAS No.: 40677-94-9 Molecular Formula: C10H5F11O2 Formula Weight: 366.13 Storage Temperature: 2°C to 8°C Physical Form: Liquid Sensitivity: Air Sensitive Appearance: Colourless MDL No.: MFCD00080614

Formel: C₁₀H₅F₁₁O₂
Lagertemperatur: Kühlschrank
MDL: MFCD00080614
CAS-Nummer: 40677-94-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR