Toluol 99,8 GC_ASSAY_METHOD, ChromAR® LC, UV, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

Synonyme: Anisen, Methylbenzol, Methylbenzen, Toluen, Retinnaphtha, Phenylmethan

4483-25EA 137.41 EUR
4483-25
Toluol 99,8 GC_ASSAY_METHOD, ChromAR® LC, UV, Macron Fine Chemicals™
Toluol

Especially intended for use as an HPLC and TLC solvent.

Formel: C₆H₅CH₃
Molecular Weight: 92.14 g/mol
Siedepunkt: 110.6 °C (1013 hPa)
Schmelzpunkt: –95 °C
Dichte: 0.867 g/cm³ (20 °C)
Flammpunkt: 4 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008512
CAS-Nummer: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use
Assay (C₆H₅CH₃ by GC) ≥99.8 %
Water (H₂O by KF, coulometric) ≤0.01 %
Titrable Acid (meq/g) ≤0.0005 %
Residue after Evaporation ≤0.0005 %
Optical Absorbance (1-cm path vs water):
Absorbance - 290 nm ≤0,3
Absorbance - 300 nm ≤0,1
Absorbance - 310 nm ≤0.05
Absorbance - 330 nm ≤0.01
Absorbance - 350 nm ≤0.004


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR