Palladium copper on alumina (1% Pd, 0,3% Cu), Kugeln 2 mm, reduziert

Lieferant: Thermo Fisher Scientific

44833.36
44833.36EA 0 EUR
44833.36
Palladium copper on alumina (1% Pd, 0,3% Cu), Kugeln 2 mm, reduziert
Palladium copper on alumina

CAS No.: 7440-05-3 MDL No.: MFCD08064572

Lagertemperatur: Raumtemperatur
MDL: MFCD08064572
CAS-Nummer: 7440-05-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR