Dichloro-bis-(tricyclohexylphosphin)-palladium(II) (14,4% Pd)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
44844.03
44844.03EA
133
EUR
44844.03
Dichloro-bis-(tricyclohexylphosphin)-palladium(II) (14,4% Pd)
Bis(tricyclohexylphosphine)palladium(II) dichloride
CAS No.: 29934-17-6
Molecular Formula: PdCl2[P(C6H12)3]2
Formula Weight: 744.24
Physical Form: Powder
MDL No.: MFCD00191830
Formel:
C₃₆H₆₆Cl₂P₂Pd Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191830 CAS-Nummer: 29934-17-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...