Barium-2-ethylhexanoat (∼17,5% Ba) in 2-Ethylhexansäure

Lieferant: Thermo Scientific
Gefahr

44847.14
44847.14EA 80.2 EUR
44847.14
Barium-2-ethylhexanoat (∼17,5% Ba) in 2-Ethylhexansäure
Barium-2-ethylhexanoat

CAS No.: 2457-01-4 Molecular Formula: C16H30BaO4 Formula Weight: 423.76 Physical Form: Viscous liquid Appearance: Yellow

Formel: C₁₆H₃₀BaO₄
Molecular Weight: 423,76 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00058696
CAS-Nummer: 2457-01-4
EINECS: 219-535-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR