2-Methyl-1-pyrroline

Lieferant: Thermo Scientific
Gefahr

44853.AC
44853.ACEA 174 EUR
44853.AC
2-Methyl-1-pyrroline
2-Methyl-1-pyrroline

CAS No.: 872-32-2 Molecular Formula: C5H9N Formula Weight: 83.13 Flash Point: 10°(50°F) Physical Form: Liquid Density: 0.878 Boiling Point: 104-105° UN No.: UN2733 MDL No.: MFCD00128806

Formel: C₅H₉N
Molecular Weight: 83,13 g/mol
Siedepunkt: 104…105 °C
Dichte: 0,878
Flammpunkt: 10 °C (50 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00128806
CAS-Nummer: 872-32-2
EINECS: 212-822-9
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR