Instrument calibration(ICAL) standard, Solution B, Specpure®
Lieferant: Thermo Fisher Scientific
44856.AE
44856.AEEA
318
EUR
44856.AE
Instrument calibration(ICAL) standard, Solution B, Specpure®
Standards
Kationischer Standards
Mehrelement-Standards
Physical Form: Liquid
UN No.: UN1789
MDL No.: MFCD08064527
Concentration:
Al 1000 µg/ml ±2%
As 1000 µg/ml ±2%
Ca 10,000 µg/ml ±2%
Cr 1000 µg/ml ±2%
K 10,000 µg/ml ±2%
Mg 10,000 µg/ml ±2%
Na 10,000 µg/ml ±2%
Sb 1000 µg/ml ±2%
Se 1000 µg/ml ±2%
V 1000 µg/ml ±2%
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...