Iron(III) isopropoxide ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Unter Argon in Ampullen verpackt
Purity: 98%
CAS No.: 14995-22-3
Molecular Formula: C9H21FeO3
Formula Weight: 233.11
Physical Form: Solid mass, Ampouled under argon
Sensitivity: Moisture Sensitive
Appearance: Dark grey
UN No.: UN3181
MDL No.: MFCD00070436
Formel:
C₉H₂₁FeO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00070436 CAS-Nummer: 14995-22-3 UN: 3181 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...