Zirkonium(IV)oxid ≥99% Metall-Basis ausgeschlossen Hf, Nano-Pulver
Lieferant: Thermo Fisher Scientific
|
Achtung
|
44886.14
44886.22
44886.36
44886.14EA
82.7
EUR
44886.14
44886.22
44886.36
Zirkonium(IV)oxid ≥99% Metall-Basis ausgeschlossen Hf, Nano-Pulver
Zirkonium(IV)oxid
Purity: 99%
CAS No.: 1314-23-4
Molecular Formula: O2Zr
Formula Weight: 123.22
Physical Form: 15-25nm APS Powder, S.A. 20-30 m2/g
Density: 5.89
Appearance: White
Odour: Odourless
Melting Point: ca 2700°
Boiling Point: ca 5000°
Merck Reference: 14,10180
MDL No.: MFCD00011310
Formel:
ZrO₂ Siedepunkt: ∼5000 °C Schmelzpunkt: ∼2700 °C Dichte: 5,89 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011310 CAS-Nummer: 1314-23-4 EINECS: 215-227-2 Merck Index: 14,10180 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...