Nickel chromium iron, Pulver -325 mesh

Lieferant: Thermo Fisher Scientific
Achtung

44888.30 44888.A1
44888.30EA 202 EUR
44888.30 44888.A1
Nickel chromium iron, Pulver -325 mesh
Nickel chromium iron

Molecular Formula: Ni:Cr:Fe; 58:27-31:7-11 wt% UN No.: 3089 MDL No.: MFCD01741927

Formel: Ni:Cr:Fe;58:27-31:7-11%
Lagertemperatur: Raumtemperatur
MDL: MFCD01741927
UN: 3089
ADR: 4.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR