Eisen(III)oxid, Pulver, NanoArc® 20-40nm APS, S.A. 30-60m2/g für die Kosmetik
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Dieisentrioxid
NanoArc® cosmetic iron oxide is an auburn material offering unique cosmetic qualitites. The small particle size results in a strong Colour development combined with high translucency for a unique glowing look.
CAS No.: 1309-37-1
Molecular Formula: Fe2O3
Formula Weight: 159.69
Physical Form: 20-40nm APS Powder, S.A. 30-60m2/g
Density: 5.24
Appearance: Red-brown
Odour: Odourless
Melting Point: 1565°
MDL No.: MFCD00011008
NanoArc® cosmetic iron oxide is an auburn material offering unique cosmetic qualitites. The small particle size results in a strong Colour development combined with high translucency for a unique glowing look
Formel:
Fe₂O₃ Molecular Weight: 159.69 g/mol Schmelzpunkt: 1565 °C Dichte: 5.24 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011008 CAS-Nummer: 1309-37-1 EINECS: 215-168-2 Merck Index: 13,04058 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...