Eisen(III)oxid, Pulver, NanoArc® 20-40 nm, magnetisch S.A. 30-60m2/g

Lieferant: Thermo Scientific
Achtung

Synonyme: Dieisentrioxid

44896.36
44896.36EA 1146 EUR
44896.36
Eisen(III)oxid, Pulver, NanoArc® 20-40 nm, magnetisch S.A. 30-60m2/g
Eisen(III)oxid

Iron(III) oxide acts as a pigment, in paints, coatings and colored concretes, used in thermalite.


Iron oxide is a low cost, nontoxic, ecofriendly catalyst for various reactions such as the oxidation of carbon monoxide, reduction of nitrogen oxides and sulfur dioxide by CO, photocatalytic splitting of water, Fischer-Tropsch synthesis of hydrocarbons, catalytic decomposition of industrial dyes, water-gas shift reactions, nucleation of diamonds, conversion of phenolic and other environmentally harmful aromatic compounds, dehydrogenation of ethylbenzene to styrene and catalytic oxidations of various organic compounds. It is used in magnetic storage devices and in the production of iron, steel and alloys. It functions as a catalyst in the chemical oxidation of hydrogen peroxide used for the treatment of contaminated water with hydrogen peroxide.


NanoArc® magnetic iron oxide is a highly magnetic material in which the crystallites are >98% gamma-phase. This material is suitable for a variety of applications requiring strong magnetic properties and narrow particle ..

Formel: Fe₂O₃
Molecular Weight: 159,69 g/mol
Schmelzpunkt: 1565 °C
Dichte: 5,24
Lagertemperatur: Raumtemperatur
MDL: MFCD00011008
CAS-Nummer: 1309-37-1
EINECS: 215-168-2
Merck Index: 13,04058

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR